D-(+)-Galactosamine hydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

ACTIAS23513G5EA 21 GBP
ACTIAS23513G5 ACTIAS23513G25
D-(+)-Galactosamine hydrochloride
D(+)-Galactosamine hydrochloride
Formula: C₆H₁₄ClNO₅
MW: 215.64 g/mol
MDL Number: MFCD00135830
CAS Number: 1772-03-8
EINECS: 217-198-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR