2',6'-Dihydroxy-4'-methylacetophenone
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,5-Dihydroxy-4-acetyltoluene
ACTIAS20784G5EA
202
GBP
ACTIAS20784G1
ACTIAS20784G5
ACTIAS20784G25
2',6'-Dihydroxy-4'-methylacetophenone
2',6'-Dihydroxy-4'-methylacetophenone
Formula:
C₉H₁₀O₃ MW: 166.1739 g/mol |
MDL Number:
MFCD06656138 CAS Number: 1634-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...