3-Methoxy-5-(trifluoromethyl)benzyl bromide ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H32821.03EA
79.7
GBP
H32821.03
H32821.06
3-Methoxy-5-(trifluoromethyl)benzyl bromide ≥97%
3-Methoxy-5-(trifluoromethyl)benzyl bromide
Formula:
C₉H₈BrF₃O MW: 269.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025414 CAS Number: 916421-00-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...