3-Methoxy-2-(trifluoromethyl)benzyl bromide ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H33160.MDEA
35.3
GBP
H33160.MD
H33160.03
3-Methoxy-2-(trifluoromethyl)benzyl bromide ≥97%
3-Methoxy-2-(trifluoromethyl)benzyl bromide
Formula:
C₉H₈BrF₃O MW: 269.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398595 CAS Number: 1261828-76-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...