2-Methyl-5-(trifluoromethoxy)benzyl bromide 96%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H33930.03EA
73.7
GBP
H33930.03
H33930.06
2-Methyl-5-(trifluoromethoxy)benzyl bromide 96%
2-Methyl-5-(trifluoromethoxy)benzyl bromide
Formula:
C₉H₈BrF₃O MW: 269.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399774 CAS Number: 1261573-60-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...