2'-Fluoro-6'-hydroxyacetophenone ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-(2-Fluoro-6-hydroxyphenyl)ethanone
H26130.03EA
119
GBP
H26130.03
2'-Fluoro-6'-hydroxyacetophenone ≥98%
2'-Fluoro-6'-hydroxyacetophenone
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01090995 CAS Number: 93339-98-1 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...