2',4'-Diethoxyacetophenone

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS21152G1EA 115 GBP
ACTIAS21152G1 ACTIAS21152G5
2',4'-Diethoxyacetophenone
2',4'-Diethoxyacetophenone
Formula: C₁₂H₁₆O₃
MW: 208.2536 g/mol
MDL Number: MFCD00238582
CAS Number: 22924-18-1
EINECS: 212-587-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR