Methyl perfluorotetradecanoate 96%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L16975.06EA
39.5
GBP
L16975.06
Methyl perfluorotetradecanoate 96%
Methyl perfluorotetradecanoate
Formula:
C₁₅H₃F₂₇O₂ MW: 728.15 g/mol Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236620 CAS Number: 203302-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...