3'-Acetamidoacetophenone
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLE62659045-100GEA
0
GBP
MOLE62659045-100G
3'-Acetamidoacetophenone
3'-Acetamidoacetophenone
Formula:
C₁₀H₁₁NO₂ MW: 177,20 |
MDL Number:
MFCD00032278 CAS Number: 7463-31-2 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...