5'-Acetamido-2'-hydroxyacetophenone
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PROOMM0128.58EA
0
GBP
PROOMM0128.58
5'-Acetamido-2'-hydroxyacetophenone
5'-Acetamido-2'-hydroxyacetophenone
Formula:
C₁₀H₁₁NO₃ |
MDL Number:
MFCD00100490 CAS Number: 7298-67-1 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...