3,5-Difluoro-2-methylbenzamide ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H33022.03EA
131
GBP
H33022.03
H33022.MD
3,5-Difluoro-2-methylbenzamide ≥97%
3,5-Difluoro-2-methylbenzamide
Formula:
C₈H₇F₂NO MW: 171.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687171 CAS Number: 1323966-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...