3-Fluoro-2-methylphenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(3-Fluoro-2-methylphenyl)boronic acid
, 3-Fluoro-2-methylbenzeneboronic acid
H52752.MDEA
0
GBP
H52752.MD
3-Fluoro-2-methylphenylboronic acid ≥98%
3-Fluoro-2-methylphenylboronic acid
Formula:
C₇H₈BFO₂ MW: 153.95 g/mol Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368679 CAS Number: 163517-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...