3-tert-Butyl-5-methylphenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-2202-1GEA
199.43
GBP
COBBBB-2202-1G
3-tert-Butyl-5-methylphenylboronic acid
3-tert-Butyl-5-methylphenylboronic acid
Formula:
C₁₁H₁₇BO₂ |
MDL Number:
MFCD03411938 CAS Number: 193905-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...