4-Nonylphenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-n-Nonylbenzeneboronic acid
COBBBB-8349-1GEA
47.48
GBP
COBBBB-8349-1G
COBBBB-8349-250M
4-Nonylphenylboronic acid
4-Nonylphenylboronic acid
Formula:
C₁₅H₂₅BO₂ MW: 248.18 g/mol |
MDL Number:
MFCD02093070 CAS Number: 256383-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...