1-Nonanesulphonic acid sodium salt
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium 1-nonanesulphonate
, Sodium nonane-1-sulphonate
MOLEM15204747EA
0
GBP
MOLEM15204747
1-Nonanesulphonic acid sodium salt
1-Nonanesulphonic acid sodium salt
Formula:
C₉H₁₉NaO₃S MW: 230.30 g/mol |
MDL Number:
MFCD00025010 CAS Number: 35192-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...