4-Chloro-7-methoxyquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS2571G1EA 46 GBP
ACTIAS2571G25 ACTIAS2571G1 ACTIAS2571G5
4-Chloro-7-methoxyquinoline
4-Chloro-7-methoxyquinoline
Formula: C₁₀H₈ClNO
MW: 193.6300 g/mol
MDL Number: MFCD06659036
CAS Number: 68500-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR