4-Aminotetrahydropyran
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tetrahydro-2H-pyran-4-amine
COBBAM-1004-5GEA
0
GBP
COBBAM-1004-5G
COBBAM-1004-1G
4-Aminotetrahydropyran
4-Aminotetrahydropyran
Formula:
C₅H₁₁NO MW: 101.1469 g/mol |
MDL Number:
MFCD02179436 CAS Number: 38041-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...