5-Nitro-6-(trifluoromethoxy)quinoline

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBHC-2416-1GEA 0 GBP
COBBHC-2416-1G
5-Nitro-6-(trifluoromethoxy)quinoline
5-Nitro-6-(trifluoromethoxy)quinoline
Formula: C₁₀H₅F₃N₂O₃
MDL Number: MFCD11855900
CAS Number: 1133115-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR