3-Methoxy-2-(trifluoromethyl)benzylamine 97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H33629.03EA
144
GBP
H33629.03
H33629.MD
3-Methoxy-2-(trifluoromethyl)benzylamine 97%
3-Methoxy-2-(trifluoromethyl)benzylamine
Formula:
C₉H₁₀F₃NO MW: 205.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398592 CAS Number: 1261647-71-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...