3-Nitrobenzylamine hydrochloride
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Nitrobenzylammonium hydrochloride
ACTIAS20479G5EA
32
GBP
ACTIAS20479G5
ACTIAS20479G100
ACTIAS20479G25
3-Nitrobenzylamine hydrochloride
3-Nitrobenzylamine hydrochloride
Formula:
C₇H₉ClN₂O₂ MW: 188.62 g/mol |
MDL Number:
MFCD00012858 CAS Number: 26177-43-5 EINECS: 247-502-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...