3-Methyl-5-(trifluoromethoxy)benzamide ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H31653.06EA
214
GBP
H31653.06
H31653.03
3-Methyl-5-(trifluoromethoxy)benzamide ≥97%
3-Methyl-5-(trifluoromethoxy)benzamide
Formula:
C₉H₈F₃NO₂ MW: 219.16 g/mol Melting Pt: 148…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD09025372 CAS Number: 916420-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...