2-Methyl-5-(trifluoromethoxy)benzamide ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H33533.03EA
56.7
GBP
H33533.03
H33533.06
2-Methyl-5-(trifluoromethoxy)benzamide ≥97%
2-Methyl-5-(trifluoromethoxy)benzamide
Formula:
C₉H₈F₃NO₂ MW: 219.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399714 CAS Number: 1261828-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...