2-(Methoxymethoxy)-5-(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxymethoxy-5-(trifluoromethyl)phenylboronic acid

COBBFA-2159-1GEA 0 GBP
COBBFA-2159-1G
2-(Methoxymethoxy)-5-(trifluoromethyl)phenylboronic acid
2-(Methoxymethoxy)-5-(trifluoromethyl)phenylboronic acid
Formula: C₉H₁₀BF₃O₄
MDL Number: MFCD11856035
CAS Number: 1256355-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR