4-Cyano-3-(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBBB-5560-1GEA 0 GBP
COBBBB-5560-1G
4-Cyano-3-(trifluoromethyl)phenylboronic acid
4-Cyano-3-(trifluoromethyl)phenylboronic acid
Formula: C₈H₅BF₃NO₂
MDL Number: MFCD10697412
CAS Number: 915299-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR