3-Chloro-4-(trifluoromethoxy)benzylamine 97%, JRD
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H31785.MDEA
34.6
GBP
H31785.MD
H31785.03
3-Chloro-4-(trifluoromethoxy)benzylamine 97%, JRD
3-Chloro-4-(trifluoromethoxy)benzylamine
Formula:
C₈H₇ClF₃NO MW: 225.60 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06212980 CAS Number: 771581-60-3 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...