2-(Chloromethyl)-5-nitrophenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-5831-1GEA
205.76
GBP
COBBBB-5831-1G
COBBBB-5831-250M
COBBBB-5831-5G
2-(Chloromethyl)-5-nitrophenylboronic acid
2-(Chloromethyl)-5-nitrophenylboronic acid
Formula:
C₇H₇BClNO₄ |
MDL Number:
MFCD12025991 CAS Number: 1217500-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...