3-(trans-2-Carboxyvinyl)phenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-2175-5GEA
0
GBP
COBBBB-2175-5G
COBBBB-2175-1G
3-(trans-2-Carboxyvinyl)phenylboronic acid
3-(trans-2-Carboxyvinyl)phenylboronic acid
Formula:
C₉H₉BO₄ |
MDL Number:
MFCD01075738 CAS Number: 216144-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...