N-Fluorobenzenesulphonimide
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACTIAS20178G100EA
0
GBP
ACTIAS20178G100
ACTIAS20178G25
N-Fluorobenzenesulphonimide
N-Fluorobenzenesulphonimide
Formula:
C₁₂H₁₀FNO₄S₂ MW: 315.34 g/mol |
MDL Number:
MFCD00144885 CAS Number: 133745-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...