N-Cyclopropyl-4-fluoro-2-nitroaniline

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBAN-1226-5GEA 0 GBP
COBBAN-1226-5G
N-Cyclopropyl-4-fluoro-2-nitroaniline
N-Cyclopropyl-4-fluoro-2-nitroaniline
Formula: C₉H₉FN₂O₂
MW: 196.18 g/mol
MDL Number: MFCD08448315
CAS Number: 887351-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR