4-Fluoro-(2-methoxycarbonyl)phenylboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Fluoro-2-(methoxycarbonyl)phenyl)boronic acid

COBBBB-2728-1GEA 0 GBP
COBBBB-2728-1G
4-Fluoro-(2-methoxycarbonyl)phenylboronic acid
4-Fluoro-(2-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197.9561 g/mol
MDL Number: MFCD07363762
CAS Number: 871329-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR