3-Fluoro-5-(methylcarbamoyl)phenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(3-Fluoro-5-(methylcarbamoyl)phenyl)boronic acid
, 3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid
H53149.03EA
294
GBP
H53149.03
3-Fluoro-5-(methylcarbamoyl)phenylboronic acid ≥98%
3-Fluoro-5-(methylcarbamoyl)phenylboronic acid
Formula:
C₈H₉BFNO₃ MW: 196.97 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07363793 CAS Number: 871332-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...