5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(5-(Dimethylcarbamoyl)-2-fluorophenyl)boronic acid
, 5-Dimethylcarbamoyl-2-fluorobenzeneboronic acid
H53045.MDEA
0
GBP
H53045.MD
5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥98%
5-(Dimethylcarbamoyl)-2-fluorophenylboronic acid
Formula:
C₉H₁₁BFNO₃ MW: 211.00 g/mol Melting Pt: 143…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235102 CAS Number: 874289-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...