4-Carboxy-3-chlorophenylboronic acid ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Carboxy-3-chlorobenzeneboronic acid
H28999.06EA
103
GBP
H28999.06
4-Carboxy-3-chlorophenylboronic acid ≥97%
4-Carboxy-3-chlorophenylboronic acid
Formula:
C₇H₆BClO₄ MW: 200.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656268 CAS Number: 136496-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...