5-Chloro-(2-ethoxycarbonyl)phenylboronic acid ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (5-Chloro-2-(ethoxycarbonyl)phenyl)boronic acid5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid

H53016.MDEA 0 GBP
H53016.MD
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid ≥98%
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid
Formula: C₉H₁₀BClO₄
MW: 228.44 g/mol
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD07363744
CAS Number: 871329-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR