4-Carboxy-2-nitrophenylboronic acid ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Carboxy-2-nitrobenzeneboronic acid
H52757.03EA
76.5
GBP
H52757.03
4-Carboxy-2-nitrophenylboronic acid ≥97%
4-Carboxy-2-nitrophenylboronic acid
Formula:
C₇H₆BNO₆ MW: 210.94 g/mol Melting Pt: 265…268 °C Storage Temperature: Ambient |
MDL Number:
MFCD00448780 CAS Number: 85107-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...