3-Carboxy-5-nitrophenylboronic acid pinacol ester ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Carboxy-5-nitrobenzeneboronic acid pinacol ester
, 3-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
H52877.MDEA
23.2
GBP
H52877.MD
3-Carboxy-5-nitrophenylboronic acid pinacol ester ≥97%
3-Carboxy-5-nitrophenylboronic acid pinacol ester
Formula:
C₁₃H₁₆BNO₆ MW: 293.08 g/mol Melting Pt: 246…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411932 CAS Number: 377780-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...