(R)-(+)-N-Benzyl-α-methylbenzylamine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-(+)-N-Benzyl-1-phenylethylamine

MOLEM29580262EA 0 GBP
MOLEM29580262
(R)-(+)-N-Benzyl-α-methylbenzylamine
(R)-(+)-N-Benzyl-α-methylbenzylamine
Formula: C₁₅H₁₇N
MW: 211,31
MDL Number: MFCD00015010
CAS Number: 38235-77-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR