4'-Acetamido-3'-bromoacetophenone ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

L07626.06EA 51.6 GBP
L07626.06
4'-Acetamido-3'-bromoacetophenone ≥98%
4'-Acetamido-3'-bromoacetophenone
Formula: C₁₀H₁₀BrNO₂
MW: 256.10 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00051781
CAS Number: 101209-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR