1,3-Dimethyl-1H-pyrazole-5-boronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(1,3-Dimethyl-1H-pyrazol-5-yl)boronic acid
COBBBB-6021-250MEA
51.44
GBP
COBBBB-6021-250M
COBBBB-6021-1G
1,3-Dimethyl-1H-pyrazole-5-boronic acid
1,3-Dimethyl-1H-pyrazole-5-boronic acid
Formula:
C₅H₉BN₂O₂ MW: 139.9500 g/mol |
MDL Number:
MFCD12407383 CAS Number: 847818-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...