6-Indoleboronic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Indole-6-boronic acid

COBBBB-2932-5GEA 0 GBP
COBBBB-2932-5G COBBBB-2932-1G
6-Indoleboronic acid
6-Indoleboronic acid
Formula: C₈H₈BNO₂
MW: 160.97 g/mol
MDL Number: MFCD03095176
CAS Number: 147621-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR