6-Indoleboronic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Indole-6-boronic acid
ACTIAS50065G1EA
66
GBP
ACTIAS50065G1
ACTIAS50065G5
ACTIAS50065G25
6-Indoleboronic acid
6-Indoleboronic acid
Formula:
C₈H₈BNO₂ MW: 160.97 g/mol |
MDL Number:
MFCD03095176 CAS Number: 147621-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...