1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(Phenylsulphonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
COBBPN-8903-1GEA
0
GBP
COBBPN-8903-1G
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester
Formula:
C₂₀H₂₂BNO₄S MW: 383.27 g/mol |
MDL Number:
MFCD08276778 CAS Number: 870717-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...