Furan-2,3-dicarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL720658-5GEA
326
GBP
SIAL720658-5G
Furan-2,3-dicarboxylic acid, Sigma-Aldrich®
Furan-2,3-dicarboxylic acid
Formula:
C₆H₄O₅ MW: 156.0930 g/mol |
MDL Number:
MFCD01860231 CAS Number: 4282-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...