4-Propoxy-2-(trifluoromethyl)phenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-6212-1GEA
0
GBP
COBBBB-6212-1G
COBBBB-6212-250M
4-Propoxy-2-(trifluoromethyl)phenylboronic acid
4-Propoxy-2-(trifluoromethyl)phenylboronic acid
Formula:
C₁₀H₁₂BF₃O₃ |
MDL Number:
MFCD15143462 CAS Number: 1186482-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...