5-Chloro-2-methylphenylboronic acid ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(5-Chloro-2-methylphenyl)boronic acid
, 5-Chloro-2-methylbenzeneboronic acid
H52994.06EA
103
GBP
H52994.06
5-Chloro-2-methylphenylboronic acid ≥97%
5-Chloro-2-methylphenylboronic acid
Formula:
C₇H₈BClO₂ MW: 170.40 g/mol Melting Pt: 162…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411939 CAS Number: 148839-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...