Acetochlor
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC10018000EA
182
GBP
EHERC10018000
Acetochlor
Acetochlor
Formula:
C₁₄H₂₀ClNO₂ |
MDL Number:
MFCD00143534 CAS Number: 34256-82-1 EINECS: 251-899-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...