4-Amino-1-methylpiperidine

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ACTIAS21038G25EA 0 GBP
ACTIAS21038G25
4-Amino-1-methylpiperidine
4-Amino-1-methylpiperidine
Formula: C₆H₁₄N₂
MW: 114.1888 g/mol
MDL Number: MFCD01570549
CAS Number: 41838-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR