4-Nitro-1H-pyrazole
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACTIAS21726G25EA
16
GBP
ACTIAS21726G25
ACTIAS21726G100
4-Nitro-1H-pyrazole
4-Nitro-1H-pyrazole
Formula:
C₃H₃N₃O₂ MW: 113.0800 g/mol |
MDL Number:
MFCD00159626 CAS Number: 2075-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...