Ethyl-2-methylnicotinate
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl 2-methylnicotinate
ACTIAS23337G25EA
88
GBP
ACTIAS23337G25
ACTIAS23337G5
Ethyl-2-methylnicotinate
Ethyl 2-methylnicotinate
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol |
MDL Number:
MFCD00006336 CAS Number: 1721-26-2 EINECS: 217-013-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...