5-Methylisophthalic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACTIAS21840G100EA
0
GBP
ACTIAS21840G100
ACTIAS21840G25
5-Methylisophthalic acid
5-Methylisophthalic acid
Formula:
C₉H₈O₄ MW: 180.16 g/mol |
MDL Number:
MFCD00013986 CAS Number: 499-49-0 EINECS: 207-881-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...